missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-chlorobenzaldehyde, 98%, Thermo Scientific Chemicals

Product Code. 15471649
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15471649 5 g 5g
15461649 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15471649 Supplier Thermo Scientific Alfa Aesar Supplier No. H64438.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

It finds its application as an intermediate in organic syntheses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its application as an intermediate in organic syntheses.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 189628-37-3
Molecular Formula C7H4BrClO
Molecular Weight (g/mol) 219.46
MDL Number MFCD08445659
InChI Key DPKKRQAEYWOISP-UHFFFAOYSA-N
PubChem CID 10608925
IUPAC Name 5-bromo-2-chlorobenzaldehyde
SMILES ClC1=C(C=O)C=C(Br)C=C1

Specifications

Color White to Yellow
Quantity 5 g
Beilstein 9757388
Synonym 2-chloro-5-bromobenzaldehyde, 5-bromo-2-chloro-benzaldehyde, benzaldehyde, 5-bromo-2-chloro, pubchem10373, acmc-1bzt6, ksc498g6h, 5-bromo-2-chloro benzaldehyde, benzaldehyde,5-bromo-2-chloro, 5-bromanyl-2-chloranyl-benzaldehyde
Solubility Information Slightly soluble in water.
Formula Weight 219.46
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 5-Bromo-2-chlorobenzaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.