missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-chloroanisole, 98%

Product Code. 10123244 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10123244 5 g Glass bottle 5g
10795905 1 g Glass bottle 1g
10326974 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10123244 Supplier Thermo Scientific Acros Supplier No. 398900050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 16817-43-9 | C7H6BrClO | 221.48 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16817-43-9
Molecular Formula C7H6BrClO
Molecular Weight (g/mol) 221.48
MDL Number MFCD00078659
InChI Key UAMVKOTWSHJOSY-UHFFFAOYSA-N
Synonym 5-bromo-2-chloroanisole, 5-bromo-2-chloroanisol, 4-chloro-3-methoxybromobenzene, 2-chloro-5-bromoanisole, 5-bromo-2-chloro-anisole, benzene, 4-bromo-1-chloro-2-methoxy, 4-bromo-1-chloro-2-methoxy-benzene, zlchem 582, pubchem2632, acmc-209dxq
PubChem CID 2769567
IUPAC Name 4-bromo-1-chloro-2-methoxybenzene
SMILES COC1=C(Cl)C=CC(Br)=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 27°C to 28°C
Color Colorless to Yellow
Density 1.6310g/mL
Boiling Point 240°C to 245°C (748.0 mmHg)
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula BrC6H3(Cl)OCH3
Quantity 5 g
Specific Gravity 1.631
Formula Weight 221.48
Percent Purity 98%
Physical Form Liquid or Low Melting Solid
Chemical Name or Material 5-Bromo-2-chloroanisole
Show More Show Less
compliance-icons
Product Identifier
  • 5-Bromo-2-chloroanisole
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.