missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-chloro-3-nitropyridine, 98%

Product Code. 15442299
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15442299 25 g 25g
15452299 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15442299 Supplier Thermo Scientific Alfa Aesar Supplier No. H64897.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 67443-38-3 | C5H2BrClN2O2 | 237.44 g/mol

5-Bromo-2-chloro-3-nitropyridine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Bromo-2-chloro-3-nitropyridine is used as pharmaceutical intermediate.

Notes
Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67443-38-3
Molecular Formula C5H2BrClN2O2
Molecular Weight (g/mol) 237.44
MDL Number MFCD00222270
InChI Key WWQQPSDIIVXFOX-UHFFFAOYSA-N
Synonym 2-chloro-3-nitro-5-bromopyridine, 5-bromo-2-chloro-3-nitro-pyridine, pyridine, 5-bromo-2-chloro-3-nitro, abbypharma ap-12-5159, zlchem 393, pubchem1166, acmc-209nz1, ksc352q2l, 5-bromo-2-chloro-3-nitropyridin, 2-chloro-5-bromo-3-nitropyridine
PubChem CID 7019412
IUPAC Name 5-bromo-2-chloro-3-nitropyridine
SMILES [O-][N+](=O)C1=CC(Br)=CN=C1Cl

Specifications

Melting Point 66°C to 68°C
Color Yellow
Quantity 25 g
UN Number UN2811
Beilstein 147551
Formula Weight 237.44
Percent Purity 98%
Physical Form Crystalline
Chemical Name or Material 5-Bromo-2-chloro-3-nitropyridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.