missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-chloro-3-methylpyridine, 98%

Product Code. 15461159
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15461159 25 g 25g
15451159 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15461159 Supplier Thermo Scientific Alfa Aesar Supplier No. H64109.14
 Restricted Item
Estimated Shipment: 24-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 29241-60-9 | C6H5BrClN | 206.467 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 29241-60-9
Molecular Formula C6H5BrClN
Molecular Weight (g/mol) 206.467
MDL Number MFCD03095093
InChI Key YTSKHJMMNFPBBZ-UHFFFAOYSA-N
Synonym 2-chloro-3-methyl-5-bromopyridine, 5-bromo-2-chloro-3-picoline, 5-bromo-2-chloro-3-methyl pyridine, 5-bromo-2-chloro-3-methyl-pyridine, pyridine, 5-bromo-2-chloro-3-methyl, 2-chloro-5-bromo-3-picoline, 2-chloro-3-methyl-5-bromo pyridine, pubchem1104, ksc495o9j
PubChem CID 285434
IUPAC Name 5-bromo-2-chloro-3-methylpyridine
SMILES CC1=CC(=CN=C1Cl)Br

Specifications

Melting Point 40°C to 44°C
Quantity 25 g
Beilstein 1562796
Formula Weight 206.47
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 5-Bromo-2-chloro-3-methylpyridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.