missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-aminopyrimidine, TRC

Product Code. 30332343
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30332343 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30332343 Supplier Toronto Research Chemicals Supplier No. TRCB6215505G
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5-Bromo-2-aminopyrimidine
CAS 7752-82-1
Molecular Formula C4 H4 Br N3
Quantity 5 g
InChI Formula InChI=1S/C4H4BrN3/c5-3-1-7-4(6)8-2-3/h1-2H,(H2,6,7,8)
SMILES Nc1ncc(Br)cn1
IUPAC Name 5-bromopyrimidin-2-amine
Molecular Weight (g/mol) 174
Formula Weight 172.9589
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.