missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2,4-dihydroxybenzoic acid, 97%

Product Code. p-7024056
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11453757

Brand: Thermo Scientific Alfa Aesar A18230.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7355-22-8 | C7H5BrO4 | 233.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7355-22-8
Molecular Formula C7H5BrO4
Molecular Weight (g/mol) 233.02
MDL Number MFCD00002452
InChI Key ZRBCISXJLHZOMS-UHFFFAOYSA-N
Synonym benzoic acid, 5-bromo-2,4-dihydroxy, 5-bromo-beta-resorcylic acid, 2,4-dihydroxy-5-bromobenzoic acid, 5-bromo-.beta.-resorcylic acid, .beta.-resorcylic acid, 5-bromo, 5-bromo-2,4-dihydroxy-benzoic acid, beta-resorcylic acid, 5-bromo, 5-bromo-2,4-dihydroxy-benzoicacid, 5-bromo-2,4-dihydroxybenzoicacid, 5-bromo-b-resorcylic acid
PubChem CID 81814
IUPAC Name 5-bromo-2,4-dihydroxybenzoic acid
SMILES OC(=O)C1=CC(Br)=C(O)C=C1O

Specifications

Melting Point ∼205°C (decomposition)
Quantity 5 g
Beilstein 2967526
Formula Weight 233.02
Percent Purity 98%
Chemical Name or Material 5-Bromo-2,4-dihydroxybenzoic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.