missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2,4-dihydroxybenzoic acid, 97%

Product Code. 11463757
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11463757 25 g 25g
11453757 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11463757 Supplier Thermo Scientific Alfa Aesar Supplier No. A18230.14
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 7355-22-8 | C7H5BrO4 | 233.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7355-22-8
Molecular Formula C7H5BrO4
Molecular Weight (g/mol) 233.02
MDL Number MFCD00002452
InChI Key ZRBCISXJLHZOMS-UHFFFAOYSA-N
Synonym benzoic acid, 5-bromo-2,4-dihydroxy, 5-bromo-beta-resorcylic acid, 2,4-dihydroxy-5-bromobenzoic acid, 5-bromo-.beta.-resorcylic acid, .beta.-resorcylic acid, 5-bromo, 5-bromo-2,4-dihydroxy-benzoic acid, beta-resorcylic acid, 5-bromo, 5-bromo-2,4-dihydroxy-benzoicacid, 5-bromo-2,4-dihydroxybenzoicacid, 5-bromo-b-resorcylic acid
PubChem CID 81814
IUPAC Name 5-bromo-2,4-dihydroxybenzoic acid
SMILES OC(=O)C1=CC(Br)=C(O)C=C1O

Specifications

Melting Point ∼205°C (decomposition)
Quantity 25 g
Beilstein 2967526
Formula Weight 233.02
Percent Purity 98%
Chemical Name or Material 5-Bromo-2,4-dihydroxybenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.