missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2,4-dichloropyrimidine, 98%, Thermo Scientific Chemicals

Product Code. 10330194 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10330194 5 g Glass bottle 5g
10582052 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10330194 Supplier Thermo Scientific Acros Supplier No. 371360050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 36082-50-5
Molecular Formula C4HBrCl2N2
Molecular Weight (g/mol) 227.88
MDL Number MFCD00127818
InChI Key SIKXIUWKPGWBBF-UHFFFAOYSA-N
Synonym 2,4-dichloro-5-bromopyrimidine, 2,4-dichloro-5-bromo pyrimidine, 5-bromo-2,4-dichloro-pyrimidine, pyrimidine, 5-bromo-2,4-dichloro, 5-bromo-2,6-dichloropyrimidine, 5-bromo-2,4-dichloro-1,3-diazine, 5-brom-2,4-dichlorpyrimidin, zlchem 380, pubchem3993, acmc-1ad68
PubChem CID 289973
IUPAC Name 5-bromo-2,4-dichloropyrimidine
SMILES C1=C(C(=NC(=N1)Cl)Cl)Br

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 29.0°C to 30.0°C
Color Colorless to Yellow
Density 1.7810g/mL
Boiling Point 128.0°C (15.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.6020 to 1.6040 (20°C, 589nm)
Quantity 5 g
Specific Gravity 1.781
Formula Weight 227.88
Percent Purity 98%
Physical Form Liquid or Low Melting Solid
Chemical Name or Material 5-Bromo-2, 4-dichloropyrimidine
Show More Show Less
compliance-icons
Product Identifier
  • 5-Bromo-2,4-dichloropyrimidine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.