missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2,3-difluoropyridine, 97%

Product Code. 15491839
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15491839 5 g 5g
15401849 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15491839 Supplier Thermo Scientific Alfa Aesar Supplier No. H64566.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 89402-44-8 | C5H2BrF2N | 193.98 g/mol

It is an important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 89402-44-8
Molecular Formula C5H2BrF2N
Molecular Weight (g/mol) 193.98
MDL Number MFCD06659525
InChI Key QVIQXJRQVOPYGI-UHFFFAOYSA-N
PubChem CID 14549654
IUPAC Name 5-bromo-2,3-difluoropyridine
SMILES FC1=CC(Br)=CN=C1F

Specifications

Color Colorless to Yellow
Quantity 5 g
Beilstein 13412561
Synonym pyridine, 5-bromo-2,3-difluoro, 2,3-difluoro-5-bromopyridine, pubchem1292, 5-bromo-2,3-difluoro-pyridine, pyridine,5-bromo-2,3-difluoro, 5-bromanyl-2,3-bis fluoranyl pyridine
Solubility Information Slightly soluble in water.
Formula Weight 193.98
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 5-Bromo-2,3-difluoropyridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.