missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2,3'-bipyridine, 95%, Thermo Scientific™

Product Code. 11307509
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11307509 1 g 1g
15413807 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11307509 Supplier Thermo Scientific Alfa Aesar Supplier No. H27880.03
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 774-53-8
Molecular Formula C10H7BrN2
Molecular Weight (g/mol) 235.08
MDL Number MFCD09037420
InChI Key ILJRIAPZWHYPMK-UHFFFAOYSA-N
Synonym 5-bromo-2,3'-bipyridine, 5-bromo-2,3' bipyridinyl, 5-bromo-2-pyridin-3-yl pyridine, 5-bromo-2,3'bipyridine, 5-bromo-2,3-bipyridine, 2,3'-bipyridine, 5-bromo, 5-bromo-2,3' bipyridine
PubChem CID 11390677
SMILES BrC1=CN=C(C=C1)C1=CN=CC=C1

Specifications

Melting Point 75°C to 78°C
Quantity 1 g
IUPAC Name 5-bromo-2,3'-bipyridine
Formula Weight 235.08
Percent Purity 95%
Chemical Name or Material 5-Bromo-2,3'-bipyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.