missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2,1,3-benzothiadiazole, 97%, Thermo Scientific™

Product Code. 10652053
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 1753-75-9
Molecular Formula C6H3BrN2S
Molecular Weight (g/mol) 215.068
MDL Number MFCD00460091
InChI Key LLCRUZDFDGTAAN-UHFFFAOYSA-N
Synonym 5-bromobenzo c 1,2,5 thiadiazole, 5-bromobenzo-2,1,3-thiadiazole, 2,1,3-benzothiadiazole,5-bromo, 5-bromobenzo 1,2,5 thiadiazole, 2,1,3-benzothiadiazole, 5-bromo, pubchem12684, acmc-209ea1, amth068, 5-bromobenz-2,1,3-thiadiazole, 5-bromo-benzo 1,2,5 thiadiazole
PubChem CID 2776295
IUPAC Name 5-bromo-2,1,3-benzothiadiazole
SMILES C1=CC2=NSN=C2C=C1Br

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 215.07
Percent Purity 97%
Chemical Name or Material 5-Bromo-2, 1, 3-benzothiadiazole
compliance-icons
Product Identifier
  • 5-Bromo-2,1,3-benzothiadiazole
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'