missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-1H-indazole, 97%, Thermo Scientific™

Product Code. 15470898
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15470898 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15470898 Supplier Thermo Scientific Alfa Aesar Supplier No. H54673.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 53857-57-1
Melting Point 123°C to 127°C
Molecular Formula C7H5BrN2
MDL Number MFCD00839493,MFCD26227374
Quantity 1 g
Synonym 5-bromoindazole, 5-bromo-2h-indazole, 1h-indazole, 5-bromo, 5-bromo-1hindazole, chembl16425, 5-brom-1h-indazol, 5-bromoazaindole, 5-bromo indazole, 5-bromo-indazole
InChI Key STVHMYNPQCLUNJ-UHFFFAOYSA-N
SMILES BrC1=CC=C2NN=CC2=C1
IUPAC Name 5-bromo-1H-indazole
Molecular Weight (g/mol) 197.04
PubChem CID 761929
Formula Weight 197.03
Percent Purity 97%
Chemical Name or Material 5-Bromo-1H-indazole
Show More Show Less
Product Title
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.