missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-1H-indazole-3-carboxylic acid, 95%, Thermo Scientific™

Product Code. 10157422
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10157422 250 mg 250mg
10357782 1 g 1g
10347304 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10157422 Supplier Thermo Scientific Maybridge Supplier No. MO07350CB
 Restricted Item
Estimated Shipment: 19-10-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 1077-94-7
Molecular Formula C8H5BrN2O2
Molecular Weight (g/mol) 241.04
MDL Number MFCD05663979
InChI Key AMJVXOOGGBPVCZ-UHFFFAOYSA-N
PubChem CID 7157358
IUPAC Name 5-bromo-1H-indazole-3-carboxylic acid
SMILES OC(=O)C1=NNC2=CC=C(Br)C=C12

Specifications

Assay Percent Range 95%
Quantity 250 mg
Synonym 5-bromo-1h-indazole-3-carboxylicacid, 5-bromoindazole-3-carboxylic acid, 5-bromo-3-indazolecarboxylic acid, 1h-indazole-3-carboxylic acid, 5-bromo, 1h-indazole-3-carboxylic acid,5-bromo, pubchem7817, acmc-2098xf, ksc173q5b, 5-bromo-3-carboxy-1h-indazole, 1h-indazole-3-carboxylicacid, 5-bromo
Formula Weight 241.04
Percent Purity 95%
Chemical Name or Material 5-Bromo-1H-indazole-3-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.