missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-1H-benzimidazole, 97%, Thermo Scientific™

Product Code. 10039602
Click to view available options
Quantity:
1 g
5 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 4887-88-1
Molecular Formula C7H5BrN2
Molecular Weight (g/mol) 197.04
MDL Number MFCD00160001
InChI Key GEDVWGDBMPJNEV-UHFFFAOYSA-N
Synonym 5-bromo-1h-benzimidazole, 5-bromo-1h-benzo d imidazole, 5-bromo-1h-1,3-benzodiazole, 5-bromobenzimidazole, 1h-benzimidazole, 5-bromo, 5-bromo-3h-1,3-benzodiazole, 6-bromobenzimidazole, 5-bromo-benzimidazole, 6-bromo benzimidazole, pubchem9618
PubChem CID 785299
SMILES BrC1=CC=C2N=CNC2=C1

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
IUPAC Name 6-bromo-1H-1,3-benzodiazole
Formula Weight 197.03
Percent Purity 97%
Chemical Name or Material 5-Bromo-1H-benzimidazole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'