missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-1-pentene, 95%

Product Code. 11904501 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
2.5 g
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
2.5g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11904501 50 g Glass bottle 50g
10309310 2.5 g Glass bottle 2.5g
10559073 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11904501 Supplier Thermo Scientific Acros Supplier No. 295170500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1119-51-3 | C5H9Br | 149.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1119-51-3
Molecular Formula C5H9Br
Molecular Weight (g/mol) 149.03
MDL Number MFCD00000264
InChI Key LPNANKDXVBMDKE-UHFFFAOYSA-N
Synonym 5-bromo-1-pentene, 1-pentene, 5-bromo, 1-bromo-4-pentene, 4-pentenyl bromide, 5-bromopentene, 5-bromo-pent-1-ene, 5-bromo-pentene, 5-bromopentene-1, 5-bromo-1-amylene, pent-4-enyl bromide
PubChem CID 70704
IUPAC Name 5-bromopent-1-ene
SMILES C=CCCCBr

Specifications

CAS Min % 94.0
CAS Max % 100.0
Density 1.2600g/mL
Boiling Point 126.0°C to 127.0°C (765.0 mmHg)
Flash Point 30°C
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula Br(CH2)3CHCH2
Refractive Index 1.4620 to 1.4650
Quantity 50 g
UN Number 1993
Specific Gravity 1.26
Formula Weight 149.03
Percent Purity 95%
Chemical Name or Material 5-Bromo-1-pentene
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 5-Bromo-1-pentene
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H335-May cause respiratory irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.