Learn More
5-Bromo-1-methyl-2-oxoindoline, 97%, Maybridge™
Brand: Maybridge CC63010DA
Code : 31
Additional Details : CAS Number : 20870-90-0 Weight : 0.05100kg Transport : UN number : 2811 Chem class : 6.1 Pack group : III
Quantity | 1g |
---|
Chemical Identifiers
20870-90-0 | |
226.073 | |
WARSUKBSFLACOI-UHFFFAOYSA-N | |
2432666 | |
CN1C(=O)CC2=C1C=CC(=C2)Br |
C9H8BrNO | |
MFCD00464061 | |
5-bromo-1-methyl-2-oxoindoline, 5-bromo-1-methylindolin-2-one, 5-bromo-1-methyl-1,3-dihydro-indol-2-one, 5-bromo-1-methyl-2,3-dihydro-1h-indol-2-one, 5-bromo-1-methyl-1,3-dihydro-2h-indol-2-one, 5-bromo-1-methyl-1h-indolin-2-one, acmc-209ffi, 5-bromo-1-methyl oxindole, 1-methyl-5-bromoindoline-2-one | |
5-bromo-1-methyl-3H-indol-2-one |
Specifications
5-Bromo-1-methyl-2-oxoindoline | |
1g | |
96.5 | |
97% | |
MFCD00464061 | |
WARSUKBSFLACOI-UHFFFAOYSA-N | |
5-bromo-1-methyl-3H-indol-2-one | |
2432666 | |
97% |
Amber Glass Bottle | |
20870-90-0 | |
100.0 | |
C9H8BrNO | |
5-bromo-1-methyl-2-oxoindoline, 5-bromo-1-methylindolin-2-one, 5-bromo-1-methyl-1,3-dihydro-indol-2-one, 5-bromo-1-methyl-2,3-dihydro-1h-indol-2-one, 5-bromo-1-methyl-1,3-dihydro-2h-indol-2-one, 5-bromo-1-methyl-1h-indolin-2-one, acmc-209ffi, 5-bromo-1-methyl oxindole, 1-methyl-5-bromoindoline-2-one | |
CN1C(=O)CC2=C1C=CC(=C2)Br | |
226.073 | |
226.07 |