missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-1-methyl-1H-indole-3-carboxylic acid, 97%, Thermo Scientific Chemicals

Catalog Number 10342020
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
400071-95-6
C10H8BrNO2
254.08
MFCD14706162
WQZLFFJMRKTCMU-UHFFFAOYSA-N
5-bromo-1-methyl-1h-indole-3-carboxylic acid, 5-bromo-1-methyl-1h-indole-3-carboxylicacid, 1h-indole-3-carboxylic acid, 5-bromo-1-methyl, ksc230c9b, 5-bromo-1-methyl-1h-indole-3-carboxylic acid;
11032413
5-bromo-1-methylindole-3-carboxylic acid
CN1C=C(C2=C1C=CC(=C2)Br)C(=O)O
400071-95-6
C10H8BrNO2
254.08
MFCD14706162
WQZLFFJMRKTCMU-UHFFFAOYSA-N
5-bromo-1-methyl-1h-indole-3-carboxylic acid, 5-bromo-1-methyl-1h-indole-3-carboxylicacid, 1h-indole-3-carboxylic acid, 5-bromo-1-methyl, ksc230c9b, 5-bromo-1-methyl-1h-indole-3-carboxylic acid;
11032413
5-bromo-1-methylindole-3-carboxylic acid
CN1C=C(C2=C1C=CC(=C2)Br)C(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 255.4°C to 255.7°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 254.08
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromo-1-methyl-1H-indole-3-carboxylic acid
Show More Show Less
compliance-icons
Product Identifier
  • 5-Bromo-1-methyl-1H-indole-3-carboxylic acid, 97%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P362-Take off contaminated clothing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'