missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-1-indanone, 97%

Product Code. 10723541 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10723541 1 g Glass bottle 1g
10628444 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10723541 Supplier Thermo Scientific Acros Supplier No. 309570010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 34598-49-7 | C9H7BrO | 211.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34598-49-7
Molecular Formula C9H7BrO
Molecular Weight (g/mol) 211.06
MDL Number MFCD00082718
InChI Key KSONICAHAPRCMV-UHFFFAOYSA-N
Synonym 5-bromo-1-indanone, 5-bromoindanone, 5-bromoindan-1-one, 5-bromo-2,3-dihydro-1h-inden-1-one, 1h-inden-1-one, 5-bromo-2,3-dihydro, 1-indanone, 5-bromo, 5-bromo-indanone, 5-bromo indanone, zlchem 204, 5-bromo-1-oxoindane
PubChem CID 520695
IUPAC Name 5-bromo-2,3-dihydroinden-1-one
SMILES C1CC(=O)C2=C1C=C(C=C2)Br

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 126.0°C to 130.0°C
Color Beige to Brown
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: insoluble
Formula Weight 211.06
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 5-Bromo-1-indanone
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.