missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-1-benzothiophene-2-carbonyl chloride, 97%, Thermo Scientific™

Product Code. 10799963
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10799963 250 mg Amber Glass Bottle 250mg
10036142 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10799963 Supplier Thermo Scientific Maybridge Supplier No. CC38802CB
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 7312-14-3
Molecular Formula C9H4BrClOS
Molecular Weight (g/mol) 275.544
MDL Number MFCD05664419
InChI Key LRYLQGGHNAOVLT-UHFFFAOYSA-N
Synonym 5-bromobenzo b thiophene-2-carbonyl chloride, benzo b thiophene-2-carbonyl chloride, 5-bromo, 5-bromo-1-benzo b thiophene-2-carbonyl chloride
PubChem CID 2795570
IUPAC Name 5-bromo-1-benzothiophene-2-carbonyl chloride
SMILES C1=CC2=C(C=C1Br)C=C(S2)C(=O)Cl

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 275.55
Percent Purity 95%
Chemical Name or Material 5-Bromo-1-benzothiophene-2-carbonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.