missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-1,2-dimethyl-1H-imidazole, ≥97%, Thermo Scientific™

Product Code. 10316574
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10316574 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10316574 Supplier Thermo Scientific Maybridge Supplier No. CC25110DA
 Restricted Item
Estimated Shipment: 10-02-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 24134-09-6
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C5H7BrN2
MDL Number MFCD06659905
Quantity 1 g
Synonym 5-bromo-1,2-dimethyl-1h-imidazole, 1h-imidazole,5-bromo-1,2-dimethyl, pubchem23641, 1,2-dimethyl-5-bromo-1h-imidazole, 1h-imidazole, 5-bromo-1,2-dimethyl
InChI Key WIRCWIXZNDCIST-UHFFFAOYSA-N
SMILES CN1C(C)=NC=C1Br
IUPAC Name 5-bromo-1,2-dimethylimidazole
Molecular Weight (g/mol) 175.03
PubChem CID 7204879
Formula Weight 175.03
Percent Purity 97%
Chemical Name or Material 5-Bromo-1, 2-dimethyl-1H-imidazole
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.