missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress 5-BrdU

Product Code. 16469447
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16469447 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16469447 Supplier MedChemExpress Supplier No. HY15910

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress 5-BrdU (BrdU) is a nucleoside analog that competes with thymidine for incorporation into DNA. 5-BrdU is commonly used in the detection of proliferating cells.

Specifications

Molecular Weight (g/mol) 307.1 g/mol
Product Type Reference compound
Content And Storage 4°C, protect from light∣In solvent : -80°C, 6 months∣-20°C, 1 month (protect from light)
Shipping Condition Room temperature
Purity 95.0%
Quantity 5 g
Solubility Information DMSO : 250 mg/mL (814.07 mM∣Need ultrasonic) ∣H2O : 2 mg/mL (6.51 mM∣ultrasonic and warming and heat to 60°C)
For Use With (Application) Cancer-programmed cell death
CAS 59-14-3
Synonym BrdU∣ 5-Bromo-2'-deoxyuridine∣ BUdR
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.