missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 5-(Boc-amino)-2-(hydroxymethyl)benzeneboronic acid hemiester, 98+%

Product Code. 15470176
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15470176 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15470176 Supplier Alfa Aesar™ Supplier No. H52890.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Applications
5-(Boc-amino)-2-(hydroxymethyl)benzeneboronic acid hemiester used as a intermediate for pharmaceutical.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from ox
TRUSTED_SUSTAINABILITY

Specifications

CAS 850568-79-5
Melting Point 134°C to 138°C
Molecular Formula C12H16BNO4
MDL Number MFCD06659853
Quantity 250 mg
Synonym 6-boc-amino-1-hydroxy-2,1-benzoxaborolane, 5-boc-amino-2-hydroxymethylphenylboronic acid, dehydrate, tert-butyl 1-hydroxy-1,3-dihydrobenzo c 1,2 oxaborol-6-yl carbamate, tert-butyl n-1-hydroxy-3h-2,1-benzoxaborol-6-yl carbamate, 6-amino-1-hydroxy-2,1-benzoxaborolane, n-boc protected, 5-amino-2-hydroxymethyl benzeneboronic acid, dehydrate, n-boc protected, 5-boc-amino-2-hydroxymethylphenylboronic acid dehydrate, tert-butyl n-1-oxidanyl-3h-2,1-benzoxaborol-6-yl carbamate, 5-boc-amino-2-hydroxymethyl benzeneboronic acid hemiester, 5-boc-amino-2-hydroxymethyl benzeneboronic acid hemiester, 98+%
InChI Key PHTYIDJNPFQYPQ-UHFFFAOYSA-N
SMILES CC(C)(C)OC(=O)NC1=CC2=C(COB2O)C=C1
IUPAC Name tert-butyl N-(1-hydroxy-3H-2,1-benzoxaborol-6-yl)carbamate
Molecular Weight (g/mol) 249.07
PubChem CID 44119625
Formula Weight 249.07
Percent Purity ≥98%
Chemical Name or Material 5-(Boc-amino)-2-(hydroxymethyl)benzeneboronic acid hemiester
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.