missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 5-Benzylsulfamoyl-2-methylbenzeneboronic acid, 97%

Product Code. 15490206
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
This item is not returnable. View return policy

Product Code. 15490206

Brand: Alfa Aesar™ H52919.MD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 871329-73-6
Melting Point 118°C to 122°C
Molecular Formula C14H16BNO4S
MDL Number MFCD07363760
Quantity 250 mg
Synonym n-benzyl 3-borono-4-methylbenzenesulfonamide, 5-n-benzylsulfamoyl-2-methylphenyl boronic acid, 5-benzylsulfamoyl-2-methylbenzeneboronic acid, 4-n-benzylsulphamoyl-2-boronotoluene, n-benzyl-3-borono-4-methylbenzenesulphonamide, 5-benzylsulfamoyl-2-methylphenylboronic acid, boronic acid,b-2-methyl-5-phenylmethyl amino sulfonyl phenyl, acmc-209qgg, n-benzyl-3-borono-4-methylbenzenesulfonamide, 4-n-benzylsulfamoyl-2-boronotoluene
InChI Key VFDVOIFZPBZUIB-UHFFFAOYSA-N
SMILES B(C1=C(C=CC(=C1)S(=O)(=O)NCC2=CC=CC=C2)C)(O)O
IUPAC Name [5-(benzylsulfamoyl)-2-methylphenyl]boronic acid
Molecular Weight (g/mol) 305.155
PubChem CID 44119420
Formula Weight 305.16
Percent Purity 97%
Chemical Name or Material 5-Benzylsulfamoyl-2-methylbenzeneboronic acid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.