missing translation for 'onlineSavingsMsg'
Learn More

5-Benzyloxytryptamine hydrochloride, 98%

Product Code. p-65591 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10635522 250 mg Glass bottle 250mg
10214180 1 g Glass bottle 1g
10696204 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10635522 Supplier Thermo Scientific Acros Supplier No. 106112500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 52055-23-9 | C17H18N2O·HCl | 302.8 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52055-23-9
Molecular Formula C17H18N2O·HCl
Molecular Weight (g/mol) 302.8
MDL Number MFCD00012685
InChI Key IUWVJCIEWSQGHH-UHFFFAOYSA-N
Synonym 5-benzyloxytryptamine hydrochloride, 5-benzyloxytryptamine hcl, 2-5-phenylmethoxy-1h-indol-3-yl ethanamine hydrochloride, cambridge id 5119598, c17h18n2o.hcl, 5-benzyloxy-tryptamine hydrochloride, 5-benzyloxy tryptamine hydrochloride, 5-benzyloxytryptamine hydrochloridyl, 3-2-aminoethyl-5-benzyloxyindole hydrochloride
PubChem CID 2828785
IUPAC Name 2-(5-phenylmethoxy-1H-indol-3-yl)ethanamine;hydrochloride
SMILES C1=CC=C(C=C1)COC2=CC3=C(C=C2)NC=C3CCN.Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 255°C to 259°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 250 mg
Formula Weight 302.8
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 5-Benzyloxytryptamine hydrochloride, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 5-Benzyloxytryptamine hydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H335-May cause respiratory irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.