missing translation for 'onlineSavingsMsg'
Learn More

5-Benzyloxytryptamine hydrochloride, 98%

Product Code. 15497967
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15497967 1 g 1g
15407977 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15497967 Supplier Thermo Scientific Alfa Aesar Supplier No. H51089.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 52055-23-9 | C17H19ClN2O | 302.802 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52055-23-9
Molecular Formula C17H19ClN2O
Molecular Weight (g/mol) 302.802
MDL Number MFCD00012685
InChI Key IUWVJCIEWSQGHH-UHFFFAOYSA-N
Synonym 5-benzyloxytryptamine hydrochloride, 5-benzyloxytryptamine hcl, 2-5-phenylmethoxy-1h-indol-3-yl ethanamine hydrochloride, cambridge id 5119598, c17h18n2o.hcl, 5-benzyloxy-tryptamine hydrochloride, 5-benzyloxy tryptamine hydrochloride, 5-benzyloxytryptamine hydrochloridyl, 3-2-aminoethyl-5-benzyloxyindole hydrochloride
PubChem CID 2828785
IUPAC Name 2-(5-phenylmethoxy-1H-indol-3-yl)ethanamine;hydrochloride
SMILES C1=CC=C(C=C1)COC2=CC3=C(C=C2)NC=C3CCN.Cl

Specifications

Melting Point 259°C to 263°C
Quantity 1 g
Sensitivity Light sensitive
Formula Weight 302.8
Percent Purity 98%
Chemical Name or Material 5-Benzyloxytryptamine hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.