missing translation for 'onlineSavingsMsg'
Learn More

5-Benzyloxytryptamine hydrochloride, 98%, Thermo Scientific Chemicals

Product Code. 10635522
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52055-23-9
Molecular Formula C17H18N2O·HCl
Molecular Weight (g/mol) 302.8
MDL Number MFCD00012685
InChI Key IUWVJCIEWSQGHH-UHFFFAOYSA-N
Synonym 5-benzyloxytryptamine hydrochloride, 5-benzyloxytryptamine hcl, 2-5-phenylmethoxy-1h-indol-3-yl ethanamine hydrochloride, cambridge id 5119598, c17h18n2o.hcl, 5-benzyloxy-tryptamine hydrochloride, 5-benzyloxy tryptamine hydrochloride, 5-benzyloxytryptamine hydrochloridyl, 3-2-aminoethyl-5-benzyloxyindole hydrochloride
PubChem CID 2828785
IUPAC Name 2-(5-phenylmethoxy-1H-indol-3-yl)ethanamine;hydrochloride
SMILES C1=CC=C(C=C1)COC2=CC3=C(C=C2)NC=C3CCN.Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 255°C to 259°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 250 mg
Formula Weight 302.8
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 5-Benzyloxytryptamine hydrochloride, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 5-Benzyloxytryptamine hydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H335-May cause respiratory irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'