missing translation for 'onlineSavingsMsg'
Learn More

5-Benzyloxyindole, 95%, Thermo Scientific Chemicals

Catalog Number 10056013
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
1215-59-4
MFCD00005676
JCQLPDZCNSVBMS-UHFFFAOYSA-N
5-benzyloxyindole, 5-benzyloxy-1h-indole, 1h-indole, 5-phenylmethoxy, benzyloxy-5 indole, indole, 5-benzyloxy, 5-benzyloxy indole, unii-yci4z02e1c
14624
5-phenylmethoxy-1H-indole
C1=CC=C(C=C1)COC2=CC3=C(C=C2)NC=C3
This item is not returnable. View return policy
1215-59-4
MFCD00005676
JCQLPDZCNSVBMS-UHFFFAOYSA-N
5-benzyloxyindole, 5-benzyloxy-1h-indole, 1h-indole, 5-phenylmethoxy, benzyloxy-5 indole, indole, 5-benzyloxy, 5-benzyloxy indole, unii-yci4z02e1c
14624
5-phenylmethoxy-1H-indole
C1=CC=C(C=C1)COC2=CC3=C(C=C2)NC=C3

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 94.0
CAS Max % 100.0
Melting Point 96°C to 102°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 6% max. Toluene (GC)
Packaging Glass bottle
Molecular Formula C15H13NO
Quantity 5 g
Solubility Information Solubility in water: practically insoluble in water. Other solubilities: soluble in chloroform and (warm) ethanol
Molecular Weight (g/mol) 223.27
Formula Weight 223.27
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 5-Benzyloxyindole, 95%
Show More Show Less
compliance-icons
Product Identifier
  • 5-Benzyloxyindole
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'