missing translation for 'onlineSavingsMsg'
Learn More

5-Benzyloxyindole, 94%, may contain up to ca 7% toluene

Product Code. 11416217
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11416217 1 g 1g
11426217 5 g 5g
11436217 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11416217 Supplier Thermo Scientific Alfa Aesar Supplier No. B25143.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1215-59-4 | C15H13NO | 223.275 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1215-59-4
Molecular Formula C15H13NO
Molecular Weight (g/mol) 223.275
MDL Number MFCD00005676
InChI Key JCQLPDZCNSVBMS-UHFFFAOYSA-N
Synonym 5-benzyloxyindole, 5-benzyloxy-1h-indole, 1h-indole, 5-phenylmethoxy, benzyloxy-5 indole, indole, 5-benzyloxy, 5-benzyloxy indole, unii-yci4z02e1c
PubChem CID 14624
IUPAC Name 5-phenylmethoxy-1H-indole
SMILES C1=CC=C(C=C1)COC2=CC3=C(C=C2)NC=C3

Specifications

Melting Point 96°C to 101°C
Quantity 1 g
Beilstein 173532
Formula Weight 223.28
Percent Purity 94%
Assay may contain up to ≈7% toluene
Chemical Name or Material 5-Benzyloxyindole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.