missing translation for 'onlineSavingsMsg'
Learn More

5-(Benzyloxy)-2-bromo-4-methylaniline, 97%, Thermo Scientific™

Product Code. p-2702325
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
This item is not returnable. View return policy

Product Code. 10334102

Brand: Thermo Scientific Maybridge MO00837DA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 499770-88-6
Molecular Formula C14H14BrNO
Molecular Weight (g/mol) 292.176
MDL Number MFCD04973043
InChI Key FEMSDHJSFUHYSH-UHFFFAOYSA-N
Synonym 5-benzyloxy-2-bromo-4-methylaniline, benzyloxybromomethylaniline, 5-benzyloxy-2-bromo-4-methyl-phenylamine
PubChem CID 7018023
IUPAC Name 2-bromo-4-methyl-5-phenylmethoxyaniline
SMILES CC1=CC(=C(C=C1OCC2=CC=CC=C2)N)Br

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 292.17
Percent Purity 97%
Chemical Name or Material 5-(Benzyloxy)-2-bromo-4-methylaniline
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.