missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ 5-BDBD
GREENER_CHOICE

Product Code. p-5545660
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18014252

Brand: Tocris Bioscience™ 3579/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent and selective P2X4 antagonist

5-BDBD is a potent and selective P2X4 receptor antagonist. Blocks P2X4-mediated currents in Chinese hamster ovary cells (IC50 = 0.50 μM). Exhibits no significant antagonist effects at other P2X receptors. Reduces long-term potentiation (LTP) in rat hippocampal slices.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 50 mg
Product Type 5-BDBD
Purity >99%
Molecular Weight (g/mol) 355.19
Inhibitors 5-BDBD

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.