missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Azacytosine (4-Amino-1,3,5-triazin-2(1H)-one), Mikromol™
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
5-Azacytosine (4-Amino-1,3,5-triazin-2(1H)-one)
Specifications
Specifications
| Chemical Name or Material | 5-Azacytosine |
| CAS | 931-86-2 |
| Analyte or Component Names | 5-Azacytosine |
| Shipping Condition | Room Temperature |
| Molecular Formula | C3 H4 N4 O |
| Certifications/Compliance | ISO 9001 |
| Synonym | 1,3,5-Triazin-2(1H)-one, 6-amino-, 1,3,5-Triazin-2(1H)-one, 4-amino- (9CI), s-Triazin-2(1H)-one, 4-amino- (8CI), s-Triazin-2-ol, 4-amino- (7CI), 2-Hydroxy-4-aminotriazine, 4-Amino-1,3,5-triazin-2-ol, 4-Amino-2-hydroxy-1,3,5-triazine, 5-Azacytosine, NSC 51100, NSC 54006, Azacitidine Related Compound A |
| InChI Formula | InChI=1S/C3H4N4O/c4-2-5-1-6-3(8)7-2/h1H,(H3,4,5,6,7,8) |
| SMILES | NC1=NC(=O)NC=N1 |
| IUPAC Name | 4-amino-1H-1,3,5-triazin-2-one |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction