missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies 5-Azacytidine

Product Code. 17130012
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
250 mg
Unit Size:
250mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17130012 50 mg 50mg
17140012 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17130012 Supplier STEMCELL Technologies Supplier No. 72012

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Epigenetic modifier; Inhibits DNA methyltransferase (DNMT)

  • 5-Azacytidine is an analog of the nucleoside cytidine which can be incorporated into DNA and RNA
  • 5-Azacytidine acts as an epigenetic modifier by incorporating into DNA where it irreversibly binds to DNA methyltransferases, thus inhibiting their activity
  • Increases reprogramming efficiency of mouse fibroblasts to induced pluripotent stem (iPS) cells by inducing full reprogramming of partially reprogrammed cells (Mikkelsen et al.)
  • Resets epigenetic memory in mouse iPS cells, in combination with Trichostatin A (Kim et al.)
  • Enhances differentiation to cardiomyocytes from human embryonic stem cells (Yoon et al.)
  • Wide range of anti-metabolic activities when tested against cultured cancer cells and an effective chemotherapeutic agent for acute myelogenous leukemia

Specifications

CAS 320-67-2
Synonym 5-AzaC; Antibiotic U 18496; Ladakamycin; Mylosar; NSC 103-627; NSC 102816; U 18496; WR 183027; Zcyd
Quantity 50 mg
Molecular Formula C8H12N4O5
Purity ≥95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.