missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress 5-Azacytidine

Product Code. 16470996
Encompass_Preferred
Change view
Click to view available options
Quantity:
200 mg
Unit Size:
200mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16470996 200 mg 200mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16470996 Supplier MedChemExpress Supplier No. HY10586

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress 5-Azacytidine (Azacitidine; 5-AzaC; Ladakamycin) is a nucleoside analogue of cytidine that specifically inhibits DNA methylation. 5-Azacytidine is incorporated into DNA to covalently trap DNA methyltransferases and contributes to reverse epigenetic changes. 5-Azacytidine induces cell autophagy.

Specifications

Molecular Weight (g/mol) 244.2 g/mol
Product Type Reference compound
Content And Storage Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Shipping Condition Room temperature
Purity 98.0%
Quantity 200 mg
Solubility Information H2O : 33.33 mg/mL (136.49 mM∣Need ultrasonic) ∣DMSO : ≥ 31 mg/mL (126.95 mM)
For Use With (Application) Cancer-programmed cell death
CAS 320-67-2
Synonym Azacitidine∣ 5-AzaC∣ Ladakamycin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.