missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Aza Cytosine, TRC
Click to view available options
Quantity:
5 g
Unit Size:
5g
Specifications
Specifications
| Chemical Name or Material | 5-Azacytosine |
| CAS | 931-86-2 |
| Molecular Formula | C3 H4 N4 O |
| Quantity | 5 g |
| Synonym | 1,3,5-Triazin-2(1H)-one, 6-amino-, 1,3,5-Triazin-2(1H)-one, 4-amino- (9CI), s-Triazin-2(1H)-one, 4-amino- (8CI), s-Triazin-2-ol, 4-amino- (7CI), 2-Hydroxy-4-aminotriazine, 4-Amino-1,3,5-triazin-2-ol, 4-Amino-2-hydroxy-1,3,5-triazine, 5-Azacytosine, NSC 51100, NSC 54006 |
| InChI Formula | InChI=1S/C3H4N4O/c4-2-5-1-6-3(8)7-2/h1H,(H3,4,5,6,7,8) |
| SMILES | NC1=NC(=O)NC=N1 |
| IUPAC Name | 4-amino-1H-1,3,5-triazin-2-one |
| Molecular Weight (g/mol) | 112.09 |
| Formula Weight | 112.0385 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction