missing translation for 'onlineSavingsMsg'
Learn More

5-Aminovaleric acid, 97%

Product Code. 10387660 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10387660 25 g 25g
10205680 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10387660 Supplier Thermo Scientific Acros Supplier No. 153910250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 660-88-8 | C5H11NO2 | 117.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 660-88-8
Molecular Formula C5H11NO2
Molecular Weight (g/mol) 117.15
MDL Number MFCD00008232
InChI Key JJMDCOVWQOJGCB-UHFFFAOYSA-N
Synonym 5-aminovaleric acid, 5-amino-pentanoic acid, homopiperidinic acid, 5-aminopentanoate, valeric acid, 5-amino, delta-amino-n-valeric acid, pentanoic acid, 5-amino, delta-aminovaleric acid, 5-amino-n-valeric acid, 5-aminovalerate
PubChem CID 138
ChEBI CHEBI:15887
IUPAC Name 5-aminopentanoic acid
SMILES C(CCN)CC(=O)O

Specifications

Melting Point 158.0°C to 161.0°C
CAS Min % 96.0
CAS Max % 100.0
Color White
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass Bottle
Linear Formula H2N(CH2)4CO2H
Beilstein 04, 418
Solubility Information solution
Quantity 25 g
Formula Weight 117.15
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 5-Aminovaleric acid, 97%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.