missing translation for 'onlineSavingsMsg'
Learn More

5-Aminouracil, 97%

Product Code. 11361587
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11361587 25 g 25g
11351587 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11361587 Supplier Thermo Scientific Alfa Aesar Supplier No. L04452.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 932-52-5 | C4H5N3O2 | 127.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 932-52-5
Molecular Formula C4H5N3O2
Molecular Weight (g/mol) 127.10
MDL Number MFCD00006025
InChI Key BISHACNKZIBDFM-UHFFFAOYSA-N
Synonym 5-aminouracil, 5-amino-2,4-dihydroxypyrimidine, uracil, 5-amino, 2,4 1h,3h-pyrimidinedione, 5-amino, 2,4-dihydroxy-5-aminopyrimidine, unii-r2o7eky9g9, r2o7eky9g9, 5-aminopyrimidine-2,4-diol, 5-amino-1,3-dihydropyrimidine-2,4-dione, wbu
PubChem CID 13611
ChEBI CHEBI:46348
IUPAC Name 5-amino-1H-pyrimidine-2,4-dione
SMILES NC1=CNC(=O)NC1=O

Specifications

Melting Point >300°C
Beilstein 127250
Quantity 25 g
Formula Weight 127.1
Percent Purity 97%
Chemical Name or Material 5-Aminouracil

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.