missing translation for 'onlineSavingsMsg'
Learn More

5-Aminosalicylic acid, 99%

Product Code. 10409640 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10409640 25 g 25g
10347210 5 g 5g
10581761 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10409640 Supplier Thermo Scientific Acros Supplier No. 134330250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 89-57-6 | C7H7NO3 | 153.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 89-57-6
Molecular Formula C7H7NO3
Molecular Weight (g/mol) 153.14
MDL Number MFCD00007877
InChI Key KBOPZPXVLCULAV-UHFFFAOYSA-N
Synonym 5-aminosalicylic acid, mesalazine, mesalamine, pentasa, claversal, asacol, canasa, salofalk, rowasa, 5-asa
PubChem CID 4075
ChEBI CHEBI:6775
IUPAC Name 5-amino-2-hydroxybenzoic acid
SMILES C1=CC(=C(C=C1N)C(=O)O)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 283.0°C
Color Beige-Brown to Gray
Loss on Drying 0.5% max. (1 g, 105°C, 3 hrs)
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Plastic Bottle
Linear Formula H2NC6H3(OH)CO2H
Quantity 25 g
Beilstein 14, 579
Merck Index 15, 5974
Solubility Information Solubility in water: 0.84g/L (20°C). Other solubilities: better soluble in hot water, soluble in hcl and diluted naoh, insoluble in ethanol
Formula Weight 153.14
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 5-Aminosalicylic acid, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.