missing translation for 'onlineSavingsMsg'
Learn More

5-Aminosalicylic acid, 95%

Product Code. 11476846
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11476846 500 g 500g
11466846 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11476846 Supplier Thermo Scientific Alfa Aesar Supplier No. B23970.36
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 89-57-6 | C7H7NO3 | 153.137 g/mol

5-Aminosalicylic acid is used in the preparation of gastrointestinal anti-inflammatory agents. It is a metabolite of sulfasalazine. It acts as a drug involved in the treatment of Crohn’s disease and ulcerative colitis. Further, it is used to make dyes and light-sensitive papers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in dimethyl sulfoxide.

Notes
Moisture, air and light sensitive. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, acid chlorides, acid anhydrides and chloroformates.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 89-57-6
Molecular Formula C7H7NO3
Molecular Weight (g/mol) 153.137
MDL Number MFCD00007877
InChI Key KBOPZPXVLCULAV-UHFFFAOYSA-N
Synonym 5-aminosalicylic acid, mesalazine, mesalamine, pentasa, claversal, asacol, canasa, salofalk, rowasa, 5-asa
PubChem CID 4075
ChEBI CHEBI:6775
IUPAC Name 5-amino-2-hydroxybenzoic acid
SMILES C1=CC(=C(C=C1N)C(=O)O)O

Specifications

Melting Point ∼280°C (decomposition)
Quantity 500 g
Beilstein 2090421
Merck Index 14,5904
Solubility Information Soluble in dimethyl sulfoxide.
Formula Weight 153.14
Percent Purity 95%
Chemical Name or Material 5-Aminosalicylic acid
compliance-icons
Product Identifier
  • 5-Aminosalicylic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 5-Aminosalicylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.