missing translation for 'onlineSavingsMsg'
Learn More

5-Aminopyridine-2-carboxylic acid, 95%

Product Code. 11416500
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11416500

Brand: Thermo Scientific Alfa Aesar H27589.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 24242-20-4 | C6H6N2O2 | 138.126 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24242-20-4
Molecular Formula C6H6N2O2
Molecular Weight (g/mol) 138.126
MDL Number MFCD02684600
InChI Key WDJARUKOMOGTHA-UHFFFAOYSA-N
Synonym 5-amino-2-pyridinecarboxylic acid, 5-aminopicolinic acid, 5-amino-2-picolinic acid, 2-pyridinecarboxylic acid, 5-amino, 5-amino-pyridine-2-carboxylic acid, 5-amino-2-pyridinecarboxylicacid, pubchem15896, 5-amino-2-carboxypyridine, acmc-209ga3, ksc201o9n
PubChem CID 4179726
IUPAC Name 5-aminopyridine-2-carboxylic acid
SMILES C1=CC(=NC=C1N)C(=O)O

Specifications

Melting Point 217°C to 219°C
Odor Odorless
Quantity 5 g
Formula Weight 138.13
Percent Purity 98%
Chemical Name or Material 5-Aminopyridine-2-carboxylic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.