missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Aminopicolinic Acid, TRC
Click to view available options
Quantity:
2.5 g
Unit Size:
2.5g
Specifications
Specifications
| Chemical Name or Material | 5-Aminopicolinic Acid |
| CAS | 24242-20-4 |
| Molecular Formula | C6 H6 N2 O2 |
| Quantity | 2.5 g |
| Synonym | 2-Pyridinecarboxylic acid, 5-amino-, Picolinic acid, 5-amino- (8CI), 5-Amino-2-pyridinecarboxylic acid, 5-Aminopicolinic acid, 5-Aminopyridine-2-carboxylic acid |
| InChI Formula | InChI=1S/C6H6N2O2/c7-4-1-2-5(6(9)10)8-3-4/h1-3H,7H2,(H,9,10) |
| SMILES | Nc1ccc(nc1)C(=O)O |
| IUPAC Name | 5-aminopyridine-2-carboxylic acid |
| Molecular Weight (g/mol) | 138.12 |
| Formula Weight | 138.0429 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction