missing translation for 'onlineSavingsMsg'
Learn More

5-Aminonaphthalene-1-sulfonic acid, tech. 85%

Product Code. 11444200
Change view
Click to view available options
Quantity:
25 g
50 g
100 g
Unit Size:
100g
25g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11444200 50 g 50g
11480283 25 g 25g
11490283 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11444200 Supplier Thermo Scientific Alfa Aesar Supplier No. A11433.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 84-89-9 | C10H9NO3S | 223.25 g/mol

5-Aminonaphthalene-1-sulfonic acid is an deriva- tizing agents in the precolumn derivatization of some chiral phenoxy acid. Cu(II) based on a dansyl derivative was synthesized with 29.2% overall yield by a seven-step synthetic procedure from the readily available starting material 5-aminonaphthalene-1-sulfonic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Aminonaphthalene-1-sulfonic acid is an deriva- tizing agents in the precolumn derivatization of some chiral phenoxy acid. Cu(II) based on a dansyl derivative was synthesized with 29.2% overall yield by a seven-step synthetic procedure from the readily available starting material 5-aminonaphthalene-1-sulfonic acid.

Solubility
Soluble in hot water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 84-89-9
Molecular Formula C10H9NO3S
Molecular Weight (g/mol) 223.25
MDL Number MFCD00014315
InChI Key DQNAQOYOSRJXFZ-UHFFFAOYSA-N
Synonym 5-amino-1-naphthalenesulfonic acid, 1-naphthylamine-5-sulfonic acid, laurent acid, 1-naphthalenesulfonic acid, 5-amino, 1-amino-5-sulfonaphthalene, laurent's acid, 5-naphthylamine-1-sulfonic acid, 1-amino-5-naphthalenesufonic acid, 1-aminonaphthalene-5-sulfonic acid, unii-w70wc365od
PubChem CID 6793
IUPAC Name 5-aminonaphthalene-1-sulfonic acid
SMILES NC1=C2C=CC=C(C2=CC=C1)S(O)(=O)=O

Specifications

Melting Point >300°C
Odor Odorless
Quantity 50 g
Beilstein 2214149
Merck Index 14,6404
Solubility Information Soluble in hot water.
Formula Weight 223.25
Percent Purity 90%
Grade Technical
Chemical Name or Material 5-Aminonaphthalene-1-sulfonic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.