missing translation for 'onlineSavingsMsg'
Learn More

5-Aminolevulinic acid hydrochloride, MedChemExpress

Product Code. 16442519
Encompass_Preferred
Click to view available options
Quantity:
100 mg
1 g
5 g
10 g
Unit Size:
100mg
10g
1g
5g
This item is not returnable. View return policy

Product Code. 16442519

Brand: Medchem Express HYN0305

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress 5-Aminolevulinic acid hydrochloride (5-ALA hydrochloride) is an intermediate in heme biosynthesis in the body and the universal precursor of tetrapyrroles.

5-Aminolevulinic acid hydrochloride (5-ALA hydrochloride) is an intermediate in heme biosynthesis in the body and the universal precursor of tetrapyrroles.

Specifications

Chemical Name or Material 5-Aminolevulinic acid hydrochloride
CAS 9-2-5451
Color White
Purity Grade Notes Research
Molecular Formula C5H10ClNO3
Quantity 1 g
Synonym 5-ALA hydrochlorideδ-Aminolevulinic acid hydrochloride5-Amino-4-oxopentanoic acid hydrochloride
Solubility Information DMSO : 100 mg/mL (596.69 mM; Need ultrasonic) ∣H2O : 16.67 mg/mL (99.47 mM; ultrasonic and warming and heat to 60°C)
SMILES O=C(O)CCC(CN)=O.[H]Cl
Molecular Weight (g/mol) 167.59
Formula Weight 167.59
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.