missing translation for 'onlineSavingsMsg'
Learn More

5-Aminoimidazole-4-carboxamide, TRC

Product Code. 30005945
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30005945 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30005945 Supplier Toronto Research Chemicals Supplier No. TRCA61169025G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 360-97-4
Molecular Formula C4 H6 N4 O
Quantity 25 g
Synonym 1H-Imidazole-4-carboxamide, 5-amino-, Imidazole-4-carboxamide, 5-amino- (8CI), 5-Amino-1H-imidazole-4-carboxamide, 4-Amino-5-carbamoylimidazole, 4-Aminoimidazole-5-carboxamide, 4-Carbamoyl-5-aminoimidazole, 4-Carboxamido-5-aminoimidazole, 5-Amino-1H-imidazole-4-carboxylic acid amide, 5-Amino-3H-imidazole-4-carboxamide, 5-Amino-4-carbamoylimidazole, 5-Aminoimidazole-4-carboxamide, AIC, AICA, Ba 2756, NSC 7784, Dacarbazine Imp. B (EP), Temozolomide Imp A (EP)
InChI Formula InChI=1S/C4H6N4O/c5-3-2(4(6)9)7-1-8-3/h1H,5H2,(H2,6,9)(H,7,8)
SMILES NC(=O)c1nc[nH]c1N
IUPAC Name 5-amino-1H-imidazole-4-carboxamide
Molecular Weight (g/mol) 126.12
Formula Weight 126.0542
Chemical Name or Material 5-Amino-1H-imidazole-4-carboxamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.