missing translation for 'onlineSavingsMsg'
Learn More

5-Aminoimidazole-4-carboxamide Hydrochloride, TRC

Product Code. 30320168
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30320168 100 g 100g
30316881 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30320168 Supplier Toronto Research Chemicals Supplier No. TRCA611691100G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5-aminoimidazole-4-carboxamide hydrochloride
CAS 72-40-2
Molecular Formula C4 H6 N4 O . Cl H
Quantity 100 g
Synonym 1H-Imidazole-4-carboxamide, 5-amino-, hydrochloride (1:1), 1H-Imidazole-4-carboxamide, 5-amino-, monohydrochloride (9CI), Imidazole-4-carboxamide, 5-amino-, monohydrochloride (8CI), 4-Amino-5-carbamoylimidazole hydrochloride, 4-Aminoimidazole-5-carboxamide hydrochloride, 5-Amino-1H-imidazole-4-carboxamide hydrochloride, 5-Aminoimidazole-4-carboxamide hydrochloride
InChI Formula InChI=1S/C4H6N4O.ClH/c5-3-2(4(6)9)7-1-8-3;/h1H,5H2,(H2,6,9)(H,7,8);1H
SMILES Cl.NC(=O)c1nc[nH]c1N
IUPAC Name 5-amino-1H-imidazole-4-carboxamide;hydrochloride
Molecular Weight (g/mol) 162.58
Formula Weight 162.0308
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.