missing translation for 'onlineSavingsMsg'
Learn More

5-Aminoimidazole-4-carboxamide Hydrochloride, TRC

Product Code. 30327735
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30327735 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30327735 Supplier Toronto Research Chemicals Supplier No. TRCA61169110G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5-aminoimidazole-4-carboxamide hydrochloride
CAS 72-40-2
Molecular Formula C4 H6 N4 O . Cl H
Quantity 10 g
Synonym 1H-Imidazole-4-carboxamide, 5-amino-, hydrochloride (1:1), 1H-Imidazole-4-carboxamide, 5-amino-, monohydrochloride (9CI), Imidazole-4-carboxamide, 5-amino-, monohydrochloride (8CI), 4-Amino-5-carbamoylimidazole hydrochloride, 4-Aminoimidazole-5-carboxamide hydrochloride, 5-Amino-1H-imidazole-4-carboxamide hydrochloride, 5-Aminoimidazole-4-carboxamide hydrochloride
InChI Formula InChI=1S/C4H6N4O.ClH/c5-3-2(4(6)9)7-1-8-3;/h1H,5H2,(H2,6,9)(H,7,8);1H
SMILES Cl.NC(=O)c1nc[nH]c1N
IUPAC Name 5-amino-1H-imidazole-4-carboxamide;hydrochloride
Molecular Weight (g/mol) 162.58
Formula Weight 162.0308
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.