missing translation for 'onlineSavingsMsg'
Learn More

5-Aminoimidazole-4-carboxamide-1-Beta-D-ribofuranoside-13C2,15N, TRC

Product Code. 30008539
Change view
Click to view available options
Quantity:
0.5 mg
5 mg
Unit Size:
0.5mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30008539 5 mg 5mg
30009000 0.5 mg 0.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30008539 Supplier Toronto Research Chemicals Supplier No. TRCA6117025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1609374-70-0
Molecular Formula 13C2 C7 H14 15N N3 O5
Molecular Weight (g/mol) 261.21
Synonym 5-Aminoimidazole-4-carboxamide-1-beta-D-ribofuranoside-13C2,15N, AICAR-13C2,15N, Acadesine-13C2,15N, 5-Amino-4-imidazolecarboxamide Riboside-13C2,15N, Arasine-13C2,15N, GP 1-110-13C2,15N, NSC 105823-13C2,15N
IUPAC Name 5-amino-1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl](4,5-^{13}C_{2},3^{15}N)imidazole-4-carboxamide
SMILES N[13c]1[13c]([15n]cn1[C@@H]2O[C@H](CO)[C@@H](O)[C@H]2O)C(=O)N

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C9H14N4O5/c10-7-4(8(11)17)12-2-13(7)9-6(16)5(15)3(1-14)18-9/h2-3,5-6,9,14-16H,1,10H2,(H2,11,17)/t3-,5-,6-,9-/m1/s1/i4+1,7+1,12+1
Formula Weight 261.1002
Percent Purity >95%
Chemical Name or Material Acadesine 13C2,15N
Grade HPLC
Product Title
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.