missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress 5-Aminofluorescein

Product Code. 16482399
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mg
1 g
Unit Size:
1g
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16482399 1 g 1g
16412316 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16482399 Supplier MedChemExpress Supplier No. HYD0022
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress 5-Aminofluorescein (5-AF) is a fluorescence marker, covalently bound to human serum albumin. The excitation wavelength is 490 nm and the emission wavelength is 515 nm.

Specifications

Content And Storage 4°C, protect from light ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (protect from light)
Product Type Dye Reagents
Quantity 1 g
Shipping Condition Room temperature
Synonym 5-AF
CAS 3326-34-9
Molecular Weight (g/mol) 347.32 g/mol
Solubility DMSO : ≥ 32 mg/mL (92.13 mM)
Target HBV
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.