missing translation for 'onlineSavingsMsg'
Learn More

5-Aminobenzene-1,3-diol hydrochloride, Technical Grade, Thermo Scientific™

Product Code. 10446262
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10446262 1 g 1g
10613203 250 mg 250mg
10274684 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10446262 Supplier Thermo Scientific Maybridge Supplier No. BTB10193DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 6318-56-5
Molecular Formula C6H8ClNO2
Molecular Weight (g/mol) 161.59
MDL Number MFCD00662897
InChI Key VNZZCDQPCQIUGG-UHFFFAOYSA-N
Synonym 5-aminobenzene-1,3-diol hydrochloride, 1,3-benzenediol, 5-amino-, hydrochloride, 5-aminoresorcinol, hydrochloride, pubchem19962, 5-aminobenzene-1,3-diol hcl, 5-aminobenzene-1,3-diol hydrochloridyl
PubChem CID 459248
SMILES Cl.NC1=CC(O)=CC(O)=C1

Specifications

Packaging Amber Glass Bottle
Linear Formula NH2C6H3(OH)2.HCl
Quantity 1 g
IUPAC Name 5-aminobenzene-1,3-diol hydrochloride
Formula Weight 161.59
Percent Purity Tech.
Grade Technical
Chemical Name or Material 5-Aminobenzene-1, 3-diol hydrochloride
compliance-icons
Product Identifier
  • 5-Aminobenzene-1,3-diol hydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 5-Aminobenzene-1,3-diol hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.