missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Amino Isoquinoline, TRC
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Specifications
Specifications
| Chemical Name or Material | 5-Aminoisoquinoline |
| CAS | 1125-60-6 |
| Molecular Formula | C9 H8 N2 |
| Quantity | 250 mg |
| InChI Formula | InChI=1S/C9H8N2/c10-9-3-1-2-7-6-11-5-4-8(7)9/h1-6H,10H2 |
| SMILES | Nc1cccc2cnccc12 |
| IUPAC Name | isoquinolin-5-amine |
| Molecular Weight (g/mol) | 144.17 |
| Formula Weight | 144.0687 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction