missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Amino-4H-1,2,4-triazole-3-thiol, tech., Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
10 g
Specifications
Packaging | Amber glass bottle |
Quantity | 10 g |
Formula Weight | 116.15 |
Chemical Name or Material | 5-Amino-4H-1, 2, 4-triazole-3-thiol |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'